Anti-CENPA Mouse Monoclonal Antibody [clone: 5A7-2E11]

Proveedor: OriGene
ORIGTA326348EA 536 EUR
ORIGTA326348
Anti-CENPA Mouse Monoclonal Antibody [clone: 5A7-2E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CENPA
Clonality: Monoclonal
Clone: 5A7-2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR