Anti-CACNLB4 Mouse Monoclonal Antibody [clone: S10-7]

Proveedor: OriGene
ORIGTA326345EA 574 EUR
ORIGTA326345
Anti-CACNLB4 Mouse Monoclonal Antibody [clone: S10-7]
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: CACNLB4
Clonality: Monoclonal
Clone: S10-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR