Anti-RPS6KA1 Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326338EA 440 EUR
ORIGTA326338
Anti-RPS6KA1 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: RPS6KA1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR