Anti-NAT1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326332EA 454 EUR
ORIGTA326332
Anti-NAT1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR