Anti-PHF11 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326331EA 510 EUR
ORIGTA326331
Anti-PHF11 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PHF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR