Anti-SGK1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326326EA 550 EUR
ORIGTA326326
Anti-SGK1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR