Anti-TBC1D9 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326324EA 547 EUR
ORIGTA326324
Anti-TBC1D9 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TBC1D9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near C-terminus
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR