Anti-GOLPH3 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326322EA 510 EUR
ORIGTA326322
Anti-GOLPH3 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: GOLPH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR