Anti-ATP2A2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326319EA 510 EUR
ORIGTA326319
Anti-ATP2A2 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ATP2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Dog, Human, , Mouse, Mouse, Pig, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR