Anti-RNF13 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326318EA 456 EUR
ORIGTA326318
Anti-RNF13 Goat Polyclonal Antibody
Type: Primary
Antigen: RNF13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR