Anti-MAP2K2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326316EA 510 EUR
ORIGTA326316
Anti-MAP2K2 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR