Anti-NOLC1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326311EA 510 EUR
ORIGTA326311
Anti-NOLC1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NOLC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR