Anti-CNTN1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA326307EA 547 EUR
ORIGTA326307
Anti-CNTN1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CNTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human, , Mouse, Mouse, Pig, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR