Anti-CASP4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326297EA 564 EUR
ORIGTA326297
Anti-CASP4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:40000

Type: Primary
Antigen: CASP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR