Anti-CAPG Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327777EA 510 EUR
ORIGTA327777
Anti-CAPG Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03?g/ml

Type: Primary
Antigen: CAPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, , Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR