Anti-DDX41 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327775EA 536 EUR
ORIGTA327775
Anti-DDX41 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: DDX41
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR