Anti-PRDX6 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327774EA 510 EUR
ORIGTA327774
Anti-PRDX6 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: PRDX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR