Anti-LDHB Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327769EA 547 EUR
ORIGTA327769
Anti-LDHB Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: LDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR