Anti-LDHA Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327768EA 510 EUR
ORIGTA327768
Anti-LDHA Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR