Anti-ATP5A1 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327766EA 547 EUR
ORIGTA327766
Anti-ATP5A1 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: ATP5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Goat
Isotype: IgG
Reactivity: Bovine, Dog, Human, , Mouse, Mouse, Pig, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR