Anti-FGB Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327763EA 513 EUR
ORIGTA327763
Anti-FGB Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: FGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR