Anti-A2M Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA327759EA 547 EUR
ORIGTA327759
Anti-A2M Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR