Anti-ACHE Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326293EA 552 EUR
ORIGTA326293
Anti-ACHE Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:5000

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR