Anti-SART1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326283EA 525 EUR
ORIGTA326283
Anti-SART1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:1000 ELISA: 1:10000

Type: Primary
Antigen: SART1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR