Anti-PIAS1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326261EA 564 EUR
ORIGTA326261
Anti-PIAS1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100 ELISA: 1:10000

Type: Primary
Antigen: PIAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR