Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]

Proveedor: OriGene
ORIGTA327754EA 610 EUR
ORIGTA327754
Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR