Anti-VIL1 Mouse Monoclonal Antibody [clone: CWWB1]

Proveedor: OriGene
ORIGTA327752EA 1140 EUR
ORIGTA327752
Anti-VIL1 Mouse Monoclonal Antibody [clone: CWWB1]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: VIL1
Clonality: Monoclonal
Clone: CWWB1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR