Anti-TPSAB1 Mouse Monoclonal Antibody [clone: G3]

Proveedor: OriGene
ORIGTA327748EA 626 EUR
ORIGTA327748
Anti-TPSAB1 Mouse Monoclonal Antibody [clone: G3]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:200-1:1000

Type: Primary
Antigen: TPSAB1
Clonality: Monoclonal
Clone: G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR