Anti-THBD Mouse Monoclonal Antibody [clone: 1009]

Proveedor: OriGene
ORIGTA327745EA 1110 EUR
ORIGTA327745
Anti-THBD Mouse Monoclonal Antibody [clone: 1009]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: THBD
Clonality: Monoclonal
Clone: 1009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR