Anti-TCL1A Mouse Monoclonal Antibody [clone: MRQ-7]

Proveedor: OriGene
ORIGTA327742EA 1160 EUR
ORIGTA327742
Anti-TCL1A Mouse Monoclonal Antibody [clone: MRQ-7]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: TCL1A
Clonality: Monoclonal
Clone: MRQ-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR