Anti-SMTN Mouse Monoclonal Antibody [clone: R4A]

Proveedor: OriGene
ORIGTA327735EA 1750 EUR
ORIGTA327735
Anti-SMTN Mouse Monoclonal Antibody [clone: R4A]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: SMTN
Clonality: Monoclonal
Clone: R4A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR