Anti-PRCC Mouse Monoclonal Antibody [clone: PN-15]

Proveedor: OriGene
ORIGTA327732EA 761 EUR
ORIGTA327732
Anti-PRCC Mouse Monoclonal Antibody [clone: PN-15]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: PRCC
Clonality: Monoclonal
Clone: PN-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR