Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]

Proveedor: OriGene
ORIGTA327730EA 571 EUR
ORIGTA327730
Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:25-1:100

Type: Primary
Antigen: PSAP
Clonality: Monoclonal
Clone: PASE/4LJ
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR