Anti-KLK3 Mouse Monoclonal Antibody [clone: ER-PR8]

Proveedor: OriGene
ORIGTA327729EA 760 EUR
ORIGTA327729
Anti-KLK3 Mouse Monoclonal Antibody [clone: ER-PR8]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: ER-PR8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR