Anti-PAX8 Mouse Monoclonal Antibody [clone: MRQ-50]

Proveedor: OriGene
ORIGTA327724EA 2130 EUR
ORIGTA327724
Anti-PAX8 Mouse Monoclonal Antibody [clone: MRQ-50]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: PAX8
Clonality: Monoclonal
Clone: MRQ-50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR