Anti-GRIA2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323556EA 552 EUR
ORIGTA323556
Anti-GRIA2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: GRIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR