Anti-HMGCR Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323545EA 506 EUR
ORIGTA323545
Anti-HMGCR Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: HMGCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR