Anti-CLCN7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324441EA 574 EUR
ORIGTA324441
Anti-CLCN7 Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CLCN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR