Anti-CREB1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324428EA 609 EUR
ORIGTA324428
Anti-CREB1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:10-50; Western Blot: 1:1000; Flow Cytometry: 1:10-50

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR