Anti-BTC Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324423EA 735 EUR
ORIGTA324423
Anti-BTC Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: BTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR