Anti-TGFBR1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323851EA 564 EUR
ORIGTA323851
Anti-TGFBR1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:5000-20000; Immunochemistry: 1:100-300

Type: Primary
Antigen: TGFBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR