Anti-CLDN5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323847EA 525 EUR
ORIGTA323847
Anti-CLDN5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-2000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CLDN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR