Anti-SHC1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323832EA 552 EUR
ORIGTA323832
Anti-SHC1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:100-200

Type: Primary
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR