Anti-PLAUR Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323491EA 564 EUR
ORIGTA323491
Anti-PLAUR Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:500-2000Immunochemistry: 1:50-200

Type: Primary
Antigen: PLAUR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR