Anti-WDR52 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324413EA 734 EUR
ORIGTA324413
Anti-WDR52 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: WDR52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR