Anti-KRBA2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324401EA 735 EUR
ORIGTA324401
Anti-KRBA2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:25; Western Blot: 1:1000

Type: Primary
Antigen: KRBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR