Anti-LRRC45 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324400EA 734 EUR
ORIGTA324400
Anti-LRRC45 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:25; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: LRRC45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR