Anti-CYP1A2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA324397EA 735 EUR
ORIGTA324397
Anti-CYP1A2 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: CYP1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR