Anti-CBX5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323817EA 466 EUR
ORIGTA323817
Anti-CBX5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:200-1000

Type: Primary
Antigen: CBX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR