Anti-CD99 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA323795EA 525 EUR
ORIGTA323795
Anti-CD99 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: CD99
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR